missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ GM632 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30172
This item is not returnable.
View return policy
Description
GM632 Polyclonal antibody specifically detects GM632 in Human samples. It is validated for Western Blot
Specifications
| GM632 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 57473 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| GM632, KIAA1196, MGC149845, MGC149846, zinc finger protein 512B | |
| between 450 and 500 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering