missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GMEB1 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97215B
This item is not returnable.
View return policy
Description
GMEB1 Polyclonal antibody specifically detects GMEB1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| GMEB1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 10691 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunofluorescence | |
| Biotin | |
| DNA-binding protein p96PIF, glucocorticoid modulatory element binding protein 1, glucocorticoid modulatory element-binding protein 1, GMEB-1, P96PIF, Parvovirus initiation factor p96, PIF p96, PIF96 | |
| Produced in rabbits immunized with E. coli-derived Human GMEB1 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction