missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GMEB2 Antibody, DyLight 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-12763V
This item is not returnable.
View return policy
Description
GMEB2 Polyclonal antibody specifically detects GMEB2 in Human samples. It is validated for Western Blot
Specifications
| GMEB2 | |
| Polyclonal | |
| Western Blot | |
| DNA-binding protein p79PIF, glucocorticoid modulatory element binding protein 2, KIAA1269PIF p79, p79, P79PIF | |
| The immunogen this antibody was made to, maps to a region between residue 480 and 530 of human Glucocorticoid Modulatory Element Binding Protein 2 using the numbering given in entry NP_036516.1 (GeneID 26205). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 26205 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction