missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GMF-beta Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
€ 275.00 - € 528.00
Specifications
| Antigen | GMF-beta |
|---|---|
| Dilution | Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:200-1:500, Knockdown Validated |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18494651
|
Novus Biologicals
NBP1-89755-25ul |
25 μL |
€ 275.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18225448
|
Novus Biologicals
NBP1-89755 |
0.1 mL |
€ 528.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GMF-beta Polyclonal specifically detects GMF-beta in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| GMF-beta | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| glia maturation factor beta, glia maturation factor, beta, GMF, GMF-beta | |
| GMFB | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.04 - 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:200-1:500, Knockdown Validated | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| P60983 | |
| 2764 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:SESLVVCDVAEDLVEKLRKFRFRKETNNAAIIMKIDKDKRLVVLDEELEGISPDELKDELPERQPRFIVYSYKYQHDDGRVSYPLCFIFSSPVGCKPEQQMMYAGSKN | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 17 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title