missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GMF-beta Mouse anti-Human, Clone: 44D8, Novus Biologicals™
Mouse Monoclonal Antibody
€ 294.00 - € 486.00
Specifications
| Antigen | GMF-beta |
|---|---|
| Clone | 44D8 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Flow Cytometry 1:10-1:1083, ELISA, Immunocytochemistry/Immunofluorescence 1:100 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18232561
|
Novus Biologicals
NBP2-59490-50UL |
50 μL |
€ 294.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18202435
|
Novus Biologicals
NBP2-59490-100UL |
100 μL |
€ 486.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
GMF-beta Monoclonal specifically detects GMF-beta in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| GMF-beta | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human | |
| glia maturation factor beta, glia maturation factor, beta, GMF, GMF-beta | |
| GMFB | |
| IgG2a κ | |
| Protein A purified |
| 44D8 | |
| Western Blot 1:1000, Flow Cytometry 1:10-1:1083, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| 2764 | |
| Recombinant human GMFB (1-142aa) purified from E. coli | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title