Learn More
Thermo Scientific™ GMP Startup, Calibration, IQOQ + PT (1 Temp Mapping) and Preventive Maintenance for Cold Storage Unit (New Product)
Description
GMP Service Plan - New Product (NP) (Part of the Regulatory Assurance Plans)
Overview
- Tailored for Cold storage equipment at the new equipment stage.
- Delivers comprehensive startup, qualification, and preventive maintenance support for GMP compliance.
Included Services
- Start-up: Ensures equipment is installed, connected, and fully operational.
- Calibration: Confirms accurate performance within specified limits and maintains ISO 9001 compliance.
- Installation qualification (IQ): Verifies installation site and conditions meet manufacturer requirements. Confirms equipment and accessories arrive undamaged and properly assembled.
- Operational qualification (OQ): Verifies installed equipment operates as intended according to manufacturer specifications, including key tests such as door ajar, power failure, and temperature alerts.
- Performance testing (PT): Custom test such as 1 temperature mapping to ensure user specifications are met.
- Preventive maintenance: Routine inspections to maintain installation-level performance. Includes inspection, cleanup, worn part replacement, performance testing, and tuning.
Additional Benefits
- Priority enhanced support commitment (Next business day) in select locations
- All-inclusive repair warranty and coverage, including spare parts.
- Dedicated account coordinator for personalized support.
- 1-hour consultation for compliance and service support guidance.
- Unlimited E-learning access.
- 50% discount on remote monitoring solutions and optional add-ons (temp map/requalification)
Customer Outcomes
- Seamless startup with validated documentation.
- Reliable performance and minimized downtime.
Specifications
Specifications
| For Use With (Equipment) | ULT Freezers |
| Product Type | Service |
| Service Offering | Preventative Maintenance |