missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GMPPA Antibody, Alexa Fluor™ 700, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97140AF700
This item is not returnable.
View return policy
Description
GMPPA Polyclonal specifically detects GMPPA in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| GMPPA | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| EC 2.7.7, EC 2.7.7.13, GDP-mannose pyrophosphorylase Amannose-1-phosphate guanyltransferase alpha, GTP-mannose-1-phosphate guanylyltransferase alpha, mannose-1-phosphate guanylyltransferase (GDP) | |
| Produced in rabbits immunized with E. coli-derived Human GMPPA fragment. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 29926 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction