missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GMPR Polyclonal Antibody

GMPR Polyclonal Antibody
Brand: Thermo Scientific PA554422
This item is not returnable.
View return policy
Description
GMPR Polyclonal Antibody for Western Blot, IHC
Specifications
| GMPR | |
| Polyclonal | |
| Unconjugated | |
| GMPR | |
| 2310004P21Rik; AV028449; GMP reductase 1; GMPR; GMPR 1; Gmpr1; guanine monophosphate reductase; guanosine 5'-monophosphate oxidoreductase 1; guanosine monophosphate reductase; guanosine monophosphate reductase 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2766 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P36959 | |
| GMPR | |
| Recombinant protein corresponding to Human GMPR. Recombinant protein control fragment (Product #RP-93148). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction