missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNA14 Monoclonal Antibody (OTI3D3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812022S
This item is not returnable.
View return policy
Description
GNA14 Monoclonal antibody specifically detects GNA14 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| GNA14 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q96JG6 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 55610 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3D3 | |
| Unconjugated | |
| Vps50 | |
| AU023208; G alpha-14; G protein subunit alpha 14; Gna14; Gna-14; G-protein subunit alpha-14; guanine nucleotide binding protein (G protein), alpha 14; guanine nucleotide binding protein, alpha 14; guanine nucleotide-binding protein 14; guanine nucleotide-binding protein subunit alpha-14; TGFB inducible early growth response 3; Tieg3 | |
| Human recombinant protein fragment corresponding to amino acids 700-964 of human CCDC132 (NP_060137) produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction