Learn More
GNB1L Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26484-1-AP-20UL
 View more versions of this product
Product Code. 18037275
  • € 145.00 / 20µL
Description
The GNB1L antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human GNB1L. This antibody recognizes human antigen. The GNB1L antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
GNB1L
450 μg/mL
Rabbit
Primary
Human
150 kDa
IgG
Western Blot, ELISA
DGCRK3, FKSG1, GNB1L, GY2, KIAA1645, WDR14, WDVCF
20 μL
54584
Store at -20°C. Stable for one year after shipment.