missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GNB2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5112430
This item is not returnable.
View return policy
Description
GNB2 Polyclonal Antibody for Western Blot, ELISA
Heterotrimeric guanine nucleotide-binding proteins (G proteins), which integrate signals between receptors and effector proteins, are composed of an alpha, a beta, and a gamma subunit. These subunits are encoded by families of related genes. This gene encodes a beta subunit. Beta subunits are important regulators of alpha subunits, as well as of certain signal transduction receptors and effectors. This gene contains a trinucleotide (CCG) repeat length polymorphism in its 5' UTR.
Specifications
| GNB2 | |
| Polyclonal | |
| Unconjugated | |
| Gnb2 | |
| G protein subunit beta 2; g protein subunit beta-2; G protein, beta-2 subunit; Gnb2; Gnb-2; guanine nucleotide binding protein (G protein), beta 2; guanine nucleotide binding protein (G protein), beta polypeptide 2; guanine nucleotide binding protein beta 2 subunit; guanine nucleotide binding protein, beta 2; guanine nucleotide binding protein, beta polypeptide 2; guanine nucleotide-binding protein G(I)/G(S)/G(T) beta subunit 2; guanine nucleotide-binding protein G(I)/G(S)/G(T) subunit beta-2; guanine nucleotide-binding protein, beta 2; guanine nucleotide-binding protein, beta-2 subunit; signal-transducing guanine nucleotide-binding regulatory protein beta subunit; transducin beta chain 2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 14693, 2783 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Western Blot | |
| 0.46 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P62879, P62880 | |
| Gnb2 | |
| Recombinant Human Guanine nucleotide-binding protein G(I)/G(S)/G(T) subunit beta-2 protein (1-340AA). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction