missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNF-7, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Description
GNF-7 is a multikinase inhibitor. GNF-7 is a Bcr-Abl inhibitor, with IC50s of 133 nM and 61 nM for Bcr-AblWT and Bcr-AblT315I, respectively. GNF-7 also possesses inhibitory activity against both ACK1 (activated CDC42 kinase 1) and GCK (germinal center kinase) with IC50s of 25 nM and 8 nM, respectively. GNF-7 can be used for the research of hematologic malignancies.
Specifications
Specifications
| Chemical Name or Material | GNF-7 |
| CAS | 839706-07-9 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C28H24F3N7O2 |
| Quantity | 5 mg |
| Solubility Information | DMSO : ≥ 33 mg/mL (60.27 mM) |
| SMILES | O=C(NC1=CC=C(C)C(N2CC3=CN=C(NC4=CC=C(C)N=C4)N=C3N(C)C2=O)=C1)C5=CC=CC(C(F)(F)F)=C5 |
| Molecular Weight (g/mol) | 547.53 |
| Formula Weight | 547.53 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction