missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GNG2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16660325 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16660325 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16660325 Supplier Invitrogen™ Supplier No. PA5103875

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total GNG2.

This gene encodes one of the gamma subunits of a guanine nucleotide-binding protein. Such proteins are involved in signaling mechanisms across membranes. Various subunits forms heterodimers which then interact with the different signal molecules.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GNG2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene GNG2
Gene Accession No. P59768, P63213
Gene Alias 82; G gamma-I; G protein subunit gamma 2; GBG2; GNG2; guanine nucleotide binding protein (G protein), gamma 2; guanine nucleotide binding protein (G protein), gamma 2 subunit; guanine nucleotide binding protein gamma 2; guanine nucleotide-binding protein G(I)/G(O) gamma-2 subunit; guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-2
Gene Symbols GNG2
Host Species Rabbit
Immunogen A synthesized peptide derived from human GNG2(Accession P59768), corresponding to amino acid residues E22-L71.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14702, 54331
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.