missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GNG3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597256
This item is not returnable.
View return policy
Description
Immunogen sequence: MKGETPVNST MSIGQARKMV EQLKIEASLC RIKVSKAAAD LMTYCDAHAC EDPLITPVPT SENPFREKKF FCALL; Positive Samples: Mouse brain, Rat brain; Cellular Location: Cell membrane, Cytoplasmic side, Lipid-anchor.
Guanine nucleotide-binding proteins (G proteins) are involved as a modulator or transducer in various transmembrane signaling systems. The beta and gamma chains are required for the GTPase activity, for replacement of GDP by GTP, and for G protein-effector interaction.
Specifications
| GNG3 | |
| Polyclonal | |
| Unconjugated | |
| GNG3 | |
| [g]3; G protein subunit gamma 3; Gng3; Gngt3; G-protein gamma 3 subunit; guanine nucleotide binding protein (G protein), gamma 3; guanine nucleotide binding protein (G protein), gamma 3 subunit; guanine nucleotide binding protein, gamma 3; guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-3; guanine nucleotide-binding protein gamma-3 subunit; guanine nucleotide-binding regulatory protein gamma-3 subunit; NBP gamma-3; wu:fj56b02; zgc:92852 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 114117, 14704 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.63 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P63216 | |
| GNG3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-75 of human GNG3 (NP_0363341). | |
| 100 μL | |
| Primary | |
| Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction