Learn More
Invitrogen™ GNG8 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5103880
Product Code. 16610335
  • € 489.00 / 100µL
Description
Antibody detects endogenous levels of total GNG8.
Heterotrimeric G proteins function to relay information from cell surface receptors to intracellular effectors. Each of a very broad range of receptors specifically detects an extracellular stimulus (a photon, pheromone, odorant, hormone or neurotransmitter) while the effectors (e.g., adenyl cyclase), which act to generate one or more intracellular messengers, are less numerous. In mammals, G protein α, β and γ polypeptides are encoded by at least 16, 4 and 7 genes, respectively. Most interest in G proteins has been focused on their α subunits, since these proteins bind and hydrolyze GTP and most obviously regulate the activity of the best studied effectors. Evidence, however, has established an important regulatory role for the βγ subunits. It is becoming increasingly clear that different G protein complexes expressed in different tissues carry structurally distinct members of the γ as well as the α and β subunits and that preferential associations between members of subunit families increase G protein functional diversity.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GNG8
Polyclonal
Unconjugated
GNG8
G protein subunit gamma 8; G(y)8; gamma-9; Ggamma 9; Ggamma9; Gng8; GNG9; GNGT9; G-protein gamma 8 subunit; guanine nucleotide binding protein (G protein), gamma 8; guanine nucleotide binding protein (G protein), gamma 8 subunit; guanine nucleotide-binding protein G(I)/G(S)/G(O) subunit gamma-8
Rabbit
Affinity chromatography
RUO
14709, 245986, 94235
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P63077, P63078, Q9UK08
GNG8
A synthesized peptide derived from human GNG8(Accession Q9UK08), corresponding to amino acid residues L36-L70.