missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GNMT Monoclonal Antibody (691305)
GREENER_CHOICE

Product Code. 15594002
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15594002 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15594002 Supplier Invitrogen™ Supplier No. MA524316

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.

The protein encoded by this gene is an enzyme that catalyzes the conversion of S-adenosyl-L-methionine to S-adenosyl-L-homocysteine and sarcosine. The encoded protein is found in the cytoplasm and acts as a homotetramer. Defects in this gene are a cause of GNMT deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GNMT
Applications Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 691305
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene GNMT
Gene Accession No. Q14749
Gene Alias 2.1.1.20; epididymis secretory sperm binding protein Li 182mP; folate-binding protein; glycine methyltransferase; glycine N methyl transferase; glycine N-methyltransferase; GNMT; HEL-S-182mP
Gene Symbols GNMT
Host Species Mouse
Immunogen E. coli-derived recombinant human Glycine N-methyltransferase/GNMT Met1-Asp295.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27232
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.