missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GNPDA2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578634
This item is not returnable.
View return policy
Description
Positive Control: 293T, A431, HeLa, HepG2 Predicted Reactivity: Mouse (100%), Rat (100%), Rhesus Monkey (100%), Bovine (100%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
GNPDA2 (Glucosamine-6-phosphate deaminase) is an allosteric enzyme that catalyzes the reversible conversion of D-glucosamine-6-phosphate into D-fructose-6-phosphate and ammonium. GNPDA2 is the second mammalian glucosamine-6-phosphate deaminase discovered, and is closer in structure and activity to the E. coli enzyme than GNPDA1. GNPDA1 possesses greater affinity for ammonium than either GNPDA2 or the E. coli enzyme suggesting that the forward reaction of D-glucosamine-6-phosphate into D-fructose-6-phosphate and ammonium is catalyzed at a slower rate than GNPDA2.
Specifications
| GNPDA2 | |
| Polyclonal | |
| Unconjugated | |
| Gnpda2 | |
| 4921523I18Rik; 4933412A11Rik; BB189630; glcN6P deaminase 2; glucosamine-6-phosphate deaminase 2; Glucosamine-6-phosphate isomerase 2; glucosamine-6-phosphate isomerase SB52; GNP2; GNPDA; GNPDA 2; GNPDA2; Kiaa4008; mKIAA4008; oscillin; Sb52 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 132789 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1.38 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| Q8TDQ7 | |
| Gnpda2 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of human GNPDA2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction