missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ GNPNAT1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15738611 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15738611 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15738611 Supplier Invitrogen™ Supplier No. PA560788

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: TPMFDPSLLK EVDWSQNTAT FSPAISPTHP GEGLVLRPLC TADLNRGFFK VLGQLTETGV VSPEQFMKSF EHMKKSGDYY VTVVEDVTLG QIVATATLII EHKFIHSCAK RGRVEDVVVS DECRGKQLGK LLLSTLTLLS KKLNCYKITL ECLPQNVGFY KKFGYTVSEE NYMCRR Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

Glucosamine-6 phosphate-N-acetyl Transferase (GNPNAT1) is a key enzyme in the hexosamine biosynthetic pathway. It converts D-glucosamine 6-phosphate to N-acetyl-D-glucosamine 6-phosphate, the acetylation step allowing carbon and nitrogen enter into the hexosamine biosynthetic pathway. Recently inhibition of GNPNAT1 has been reported to promote proliferation and aggressiveness of castration-resistant prostate cancer. (27194471).
TRUSTED_SUSTAINABILITY

Specifications

Antigen GNPNAT1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene GNPNAT1
Gene Accession No. Q96EK6, Q9JK38
Gene Alias AU017428; AU040593; EMeg32; Glucosamine 6-phosphate N-acetyltransferase; glucosamine-6-phosphate acetyltransferase; glucosamine-phosphate N-acetyltransferase 1; glucosamine-phosphate N-acetyltransferase 1; glucosamine-6-phosphate acetyltransferase; glucosamine-phosphate N-acetyltransferase; GNA1; GNPNAT; GNPNAT1; Gpnat1; Gsnpat; Phosphoglucosamine acetylase; phosphoglucosamine transacetylase; Protein EMeg32; RGD1563144
Gene Symbols GNPNAT1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human GNPNAT1. Recombinant protein control fragment (Product #RP-88921).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 498486, 54342, 64841
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.