missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNPTG Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97807AF594
This item is not returnable.
View return policy
Description
GNPTG Polyclonal specifically detects GNPTG in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| GNPTG | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| C16orf27, c316G12.3, CAB56184, GlcNAc-1-phosphotransferase subunit gamma, GNPTAGchromosome 16 open reading frame 27, LP2537, N-acetylglucosamine-1-phosphate transferase, gamma subunit, N-acetylglucosamine-1-phosphotransferase subunit gamma, N-acetylglucosamine-1-phosphotransferase, gamma subunit, RJD9, UDP-N-acetylglucosamine-1-phosphotransferase subunit gamma | |
| Produced in rabbits immunized with E. coli-derived Human GNPTG fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 84572 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction