missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GnRHR Antibody (A9E4), PE , Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11601PE
This item is not returnable.
View return policy
Description
GnRHR Monoclonal antibody specifically detects GnRHR in Human samples. It is validated for Flow Cytometry
Specifications
| GnRHR | |
| Monoclonal | |
| PE | |
| GnRH receptor, GnRH-R, GNRHR1, gonadotropin-releasing hormone (type 1) receptor 1, gonadotropin-releasing hormone receptor, GRHR, leutinizing-releasing hormone receptor, LHRHRleutinizing hormone releasing horomone receptor, LRHR, luliberin receptor, type I GnRH receptor | |
| A synthetic peptide aa 1-29 (MANSASPEQNQHCSAINNSIPLMQGNLPY) from the N-terminal of human GnRHR. | |
| 0.1 mL | |
| GPCR | |
| 2798 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry | |
| A9E4 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction