Learn More
ABCD4, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of ABCD4.
Brand: Abnova PAB11482.100ug
Description
The protein encoded by this gene is a member of the superfamily of ATP-binding cassette (ABC) transporters. ABC proteins transport various molecules across extra- and intra-cellular membranes. ABC genes are divided into seven distinct subfamilies (ABC1, MDR/TAP, MRP, ALD, OABP, GCN20, White). This protein is a member of the ALD subfamily, which is involved in peroxisomal import of fatty acids and/or fatty acyl-CoAs in the organelle. All known peroxisomal ABC transporters are half transporters which require a partner half transporter molecule to form a functional homodimeric or heterodimeric transporter. The function of this peroxisomal membrane protein is unknown. However, it is speculated that it may function as a heterodimer for another peroxisomal ABC transporter and, therefore, may modify the adrenoleukodystrophy phenotype. It may also play a role in the process of peroxisome biogenesis. Alternative splicing results in at least two different transcript variants, one which is protein-coding and one which is probably not protein-coding. [provided by RefSeq
Sequence: C-RDDIDNPDQRISQDSpecifications
| ABCD4 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:8000) Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| ABCD4 | |
| ABC41/EST352188/P70R/P79R/PMP69/PXMP1L | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 5826 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of ABCD4. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_005041.1 | |
| ABCD4 | |
| A synthetic peptide corresponding to human ABCD4. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only