Learn More
AHSG, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against a recombinant AHSG.
Brand: Abnova PAB10313.500ug
Description
Alpha2-HS glycoprotein (AHSG), a glycoprotein present in the serum, is synthesized by hepatocytes. The AHSG molecule consists of two polypeptide chains, which are both cleaved from a proprotein encoded from a single mRNA. It is involved in several functions, such as endocytosis, brain development and the formation of bone tissue. The protein is commonly present in the cortical plate of the immature cerebral cortex and bone marrow hemopoietic matrix, and it has therefore been postulated that it participates in the development of the tissues. However, its exact significance is still obscure. [provided by RefSeq
Specifications
| AHSG | |
| Polyclonal | |
| Goat polyclonal antibody raised against a recombinant AHSG. | |
| Lyophilized from 20mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| A2HS/AHS/FETUA/HSGA | |
| Goat | |
| 500 μg | |
| Primary | |
| Human | |
| Lyophilized |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:40000-1:200000) Western Blot (1:10000-1:20000) The optimal working dilution should be determined by the end user. | |
| AHSG | |
| AHSG | |
| Recombinant protein corresponding to human AHSG. | |
| RUO | |
| 197 | |
| Store at 4°C on dry atmosphere. After reconstitution with deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only