Learn More
AMID, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of AMID.
Brand: Abnova PAB6734.100ug
Description
The protein encoded by this gene has significant homology to NADH oxidoreductases and the apoptosis-inducing factor PDCD8/AIF. Overexpression of this gene has been shown to induce apoptosis. The expression of this gene is found to be induced by tumor suppressor protein p53 in colon caner cells. [provided by RefSeq
Sequence: GSQVSVESGALHCSpecifications
| AMID | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:128000) The optimal working dilution should be determined by the end user. | |
| AIFM2 | |
| AMID/PRG3/RP11-367H5.2 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 84883 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of AMID. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_116186 | |
| AIFM2 | |
| A synthetic peptide corresponding to human AMID. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only