Learn More
ARL2, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of ARL2.
Brand: Abnova PAB6216.100ug
Description
This gene encodes a small GTP-binding protein of the RAS superfamily which functions as an ADP-ribosylation factor (ARF). The encoded protein is one of a functionally distinct group of ARF-like genes. [provided by RefSeq
Sequence: C-LDDISSRIFTADSpecifications
| ARL2 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:1000) Western blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| ARL2 | |
| ARFL2 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 402 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of ARL2. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_001658 | |
| ARL2 | |
| A synthetic peptide corresponding to human ARL2. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only