Learn More
BAT3, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised aganist synthetic peptide of BAT3.
Brand: Abnova PAB15612.100ug
Description
This gene was first characterized as part of a cluster of genes located within the human major histocompatibility complex class III region. This gene encodes a nuclear protein that is cleaved by caspase 3 and is implicated in the control of apoptosis. In addition, the protein forms a complex with E1A binding protein p300 and is required for the acetylation of p53 in response to DNA damage. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: C-PETNAPNHPSPAEYSpecifications
| BAT3 | |
| Polyclonal | |
| Unconjugated | |
| Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections)(3-5 ug/mL) ELISA (1:128000) The optimal working dilution should be determined by the end user. | |
| BAT3 | |
| BAG-6/BAG6/D6S52E/G3 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 7917 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed) | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised aganist synthetic peptide of BAT3. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_004630.3 | |
| BAT3 | |
| A synthetic peptide corresponding to amino acids at internal region of human BAT3. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only