missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BLK, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of BLK.
Brand: Abnova PAB19716.100ug
This item is not returnable.
View return policy
Description
Sequence: QWSPLKVSAQDKD-CSpecifications
| BLK | |
| Polyclonal | |
| Goat polyclonal antibody raised against synthetic peptide of BLK. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_001706.2 | |
| BLK | |
| A synthetic peptide corresponding to N-terminus of human BLK. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:128000) Western Blot (0.1-0.3 ug/mL) The optimal working dilution should be determined by the end user. | |
| BLK | |
| MGC10442 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 640 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction