missing translation for 'onlineSavingsMsg'
Learn More

Goat anti-Bovine IgG (H+L) Secondary Antibody, FITC, Novus Biologicals™

Product Code. 18526172 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
2 mg
Unit Size:
2mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18526172 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18526172 Supplier Novus Biologicals Supplier No. NBP174712

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Bovine samples. It is validated for Flow Cytometry, ELISA, Immunocytochemistry/ Immunofluorescence, Immunomicroscopy
TRUSTED_SUSTAINABILITY

Specifications

Antigen IgG (H+L)
Applications Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunomicroscopy
Classification Polyclonal
Concentration 1.5 mg/mL
Conjugate FITC
Dilution Flow Cytometry 1:20-1:2000, ELISA, Immunocytochemistry/Immunofluorescence, Immunomicroscopy 1:20-1:2000
Formulation 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.8, 1 % (w/v) BSA, Protease/IgG free
Host Species Goat
Immunogen This Goat anti-Bovine IgG (H+L) Secondary Antibody [FITC] was developed against purified bovine IgG (H&L).
Purification Method Affinity purified
Quantity 2 mg
Regulatory Status RUO
Research Discipline Adaptive Immunity, Immunology
Primary or Secondary Secondary
Target Species Bovine
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.