missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C18orf8, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of C18orf8.
Brand: Abnova PAB7281.100ug
This item is not returnable.
View return policy
Description
Sequence: C-DKLNHEYKKYLDAEQSpecifications
| C18orf8 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:32000) Western blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| C18orf8 | |
| FLJ23453/HsT2591/MIC1 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 29919 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of C18orf8. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_0.37458.3 | |
| C18orf8 | |
| A synthetic peptide corresponding to internal region of human C18orf8. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur