missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C20orf20, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of C20orf20.
Brand: Abnova PAB27530.100ug
This item is not returnable.
View return policy
Description
Sequence: CIRDKFSQNIGRQVPSpecifications
| C20orf20 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:32000) Western Blot (0.3-1ug/ml) The optimal working dilution should be determined by the end user. | |
| C20orf20 | |
| Eaf7/FLJ10914/MRG15BP/MRGBP/URCC4 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 55257 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of C20orf20. | |
| In Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
| NP_060740.1 | |
| C20orf20 | |
| A synthetic peptide corresponding to internal region of human C20orf20. | |
| 100 μg | |
| Primary | |
| Chicken, Human, Mouse, Rat |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction