missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Canine IgG (H+L) Secondary Antibody, Biotin, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-69372
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Canine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Dot Blot
Specifications
| IgG (H+L) | |
| Polyclonal | |
| Biotin | |
| Lyophilized from PBS (pH 7.2) | |
| Purified normal IgG isolated from pooled dog serum. Freund's complete adjuvant is used in the first step of the immunization procedure. | |
| 1 mL | |
| Adaptive Immunity, Immunology | |
| Reconstitute by by adding 1 mL sterile distilled water, spun down to remove insoluble particles, divided into small aliquots, frozen and stored at or below -20C. | |
| Store at 4°C in the dark. |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| LYOPH | |
| Western Blot, ELISA 1:100-1:2000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin, Dot Blot | |
| Goat | |
| Ion exchange chromatography | |
| RUO | |
| Secondary | |
| Canine | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction