missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Canine IgG (H+L) Secondary Antibody, PE, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-60652PE
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Canine samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunomicroscopy
Specifications
| IgG (H+L) | |
| Polyclonal | |
| PBS | |
| This Goat anti-Canine IgG (H+L) Secondary Antibody was developed against purified canine IgG (H&L). | |
| 0.5 mL | |
| Adaptive Immunity, Immunology | |
| Canine | |
| Purified |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunomicroscopy | |
| PE | |
| Goat | |
| Affinity purified | |
| RUO | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction