Learn More
Goat anti-Canine IgG (H+L) Secondary Antibody, PerCP, Novus Biologicals™
Goat Polyclonal Antibody
Brand:  Novus Biologicals NBP1-72694PCP
Product Code. 30227783
  • € 366.00 / 0.50mL
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Canine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Dot Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG (H+L)
Polyclonal
PBS
Dog IgG whole molecule
0.5 mL
Adaptive Immunity, Immunology
Canine
IgG
Western Blot, ELISA, Immunohistochemistry, Dot Blot
PerCP
Goat
Multi-step
RUO
Secondary
Store at 4°C in the dark.