Learn More
CARD6, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of CARD6.
Brand: Abnova PAB6377.100ug
Description
This gene encodes a protein that contains a caspase recruitment domain (CARD), an antiparallel six-helical bundle that mediates homotypic protein-protein interactions. The encoded protein is a microtubule-associated protein that has been shown to interact with receptor-interacting protein kinases and positively modulate signal transduction pathways converging on activation of the inducible transcription factor NF-kB. [provided by RefSeq
Sequence: ATESTPSEIIERER-CSpecifications
| CARD6 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:32000) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5-10 ug/mL) The optimal working dilution should be determined by the end user. | |
| CARD6 | |
| CINCIN1 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 84674 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed) | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of CARD6. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_115976 | |
| CARD6 | |
| A synthetic peptide corresponding to human CARD6. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only