missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD28, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of CD28.
Brand: Abnova PAB19640.100ug
This item is not returnable.
View return policy
Description
CD28 costimulation is essential for CD4 (MIM 186940)-positive T-cell proliferation, survival, interleukin-2 (IL2; MIM 147680) production, and T-helper type-2 (Th2) development.[supplied by OMIM
Sequence: SQQLQVYSKTGFNCDSpecifications
| CD28 | |
| Polyclonal | |
| Goat polyclonal antibody raised against synthetic peptide of CD28. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_006130.1 | |
| CD28 | |
| A synthetic peptide corresponding to internal region of human CD28. | |
| 100 μg | |
| Primary | |
| The immunizing peptide corresponds to a part of the extra-cellular domain. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| Unconjugated | |
| ELISA (1:64000) Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| CD28 | |
| MGC138290/Tp44 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 940 | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction