Learn More
Goat anti-Chicken IgM Fc Secondary Antibody, Novus Biologicals™
Product Code. 18663598 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18663598 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18663598 Supplier Novus Biologicals Supplier No. NBP269676
Goat Polyclonal Antibody
IgM Fc Polyclonal antibody specifically detects IgM Fc in Chicken samples. It is validated for ELISA, Immunoprecipitation, Immunoelectrophoresis
TRUSTED_SUSTAINABILITY
Specifications
Antigen IgM Fc
Applications ELISA, Immunoprecipitation, Immunoelectrophoresis
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution ELISA, Immunoprecipitation 1:10 - 1:500, Immunoelectrophoresis
Formulation Whole antiserum
Host Species Goat
Immunogen Purified IgM isolated from chicken serum. Freund's complete adjuvant is used in the first step of the immunization procedure.
Purification Method Whole Antiserum
Quantity 1 mL
Regulatory Status RUO
Research Discipline B Cell Development and Differentiation Markers, Immunology
Primary or Secondary Secondary
Reconstitution Reconstitute the lyophilized antiserum by adding 1 mL sterile distilled water. Dilutions may be prepared by adding phosphate buffered saline (PBS, pH 7.2).
Target Species Chicken
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Lyophilized