Learn More
Goat anti-Chicken IgM Heavy Chain Secondary Antibody, HRP, Novus Biologicals™
Goat Polyclonal Antibody
Brand:  Novus Biologicals NBP2-60690H
Product Code. 30232209
  • € 432.00 / 0.50mL
Description
IgM Heavy Chain Polyclonal antibody specifically detects IgM Heavy Chain in Chicken samples. It is validated for Western Blot, ELISA, Immunohistochemistry, SDS-Page
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgM Heavy Chain
Polyclonal
PBS
Goat
Multi-step
RUO
Secondary
Chicken
Purified
Western Blot, ELISA, Immunohistochemistry, SDS-Page
HRP
Constant region of heavy chain of IgM, Ig Mu Chain C Region BOT, Ig Mu Chain C Region GAL, Ig Mu Chain C Region OU, immunoglobulin heavy constant mu, Immunoglobulin mu chain
Chicken IgM whole molecule
0.5 mL
B Cell Development and Differentiation Markers, Immunology
3507
Store at 4°C in the dark.