missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Chicken IgY Fc Secondary Antibody, Alkaline Phosphatase, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB7287-0.5mg
This item is not returnable.
View return policy
Description
IgY Fc Polyclonal antibody specifically detects IgY Fc in Chicken samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| IgY Fc | |
| Polyclonal | |
| Alkaline Phosphatase (AP) | |
| 50 mM HEPES pH 7.1, 0.1 M NaCl, 1 mM MgCl2, 0.1 mM ZnCl2 containing 0.2%BSA | |
| Chicken IgG (IgY)-Fc Fragment | |
| 0.5 mg | |
| Secondary | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 1:5000 - 1:50000, ELISA 1:500 to 1:5 000, Immunohistochemistry 1:100- 1:1000, Immunocytochemistry/Immunofluorescence 1:100- 1:1000, Immunohistochemistry-Paraffin 1:100- 1:1000 | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Chicken | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction