missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Chicken IgY (H+L) Secondary Antibody, HRP (Pre-adsorbed), Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB7303
This item is not returnable.
View return policy
Description
IgY (H+L) Polyclonal specifically detects IgY (H+L) in Chicken samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| IgY (H+L) | |
| Polyclonal | |
| HRP | |
| Goat | |
| Affinity Purified | |
| RUO | |
| By immunoelectrophoresis and ELISA this antibody reacts specifically with chicken IgG and with light chains common to other chicken immunoglobulins. No antibody was detected against nonimmunoglobulin serum proteins. Antiserum was cross adsorbed using bovine, horse, human, mouse, pig, rabbit, and rat immunosorbents to remove cross reactive Antibodies. | |
| Store at 4C. Do not freeze. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 1:1000- 1:30000, ELISA 1:10000-1:100000, Immunohistochemistry 1:200 - 1:5000, Immunocytochemistry/Immunofluorescence 1:200 - 1:5000, Immunohistochemistry-Paraffin 1:200-1:5000 | |
| Chicken IgG (IgY)-heavy and light chain | |
| 1 mL | |
| Secondary | |
| Chicken | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction