Learn More
CKB, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of CKB.
Brand: Abnova PAB6995.100ug
Description
The protein encoded by this gene is a cytoplasmic enzyme involved in energy homeostasis. The encoded protein reversibly catalyzes the transfer of phosphate between ATP and various phosphogens such as creatine phosphate. It acts as a homodimer in brain as well as in other tissues, and as a heterodimer with a similar muscle isozyme in heart. The encoded protein is a member of the ATP:guanido phosphotransferase protein family. A pseudogene of this gene has been characterized. [provided by RefSeq
Sequence: C-KPSDEHKTDLNPDNSpecifications
| CKB | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:2000) Western Blot (0.1-0.3 ug/mL) The optimal working dilution should be determined by the end user. | |
| CKB | |
| B-CK/CKBB | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 1152 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of CKB. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_001814.2 | |
| CKB | |
| A synthetic peptide corresponding to human CKB. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only