Learn More
CLEC4C, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of CLEC4C.
Brand: Abnova PAB17024.100ug
Description
This gene encodes a member of the C-type lectin/C-type lectin-like domain (CTL/CTLD) superfamily. Members of this family share a common protein fold and have diverse functions, such as cell adhesion, cell-cell signalling, glycoprotein turnover, and roles in inflammation and immune response. The encoded type 2 transmembrane protein may play a role in dendritic cell function. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq
Sequence: QSWTKSQKNCSVSpecifications
| CLEC4C | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:8000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| CLEC4C | |
| BDCA2/CD303/CLECSF11/CLECSF7/DLEC/HECL/MGC125789/MGC125791/MGC125792/MGC125793/PRO34150 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 170482 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of CLEC4C. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_569708.1;NP_987099.1 | |
| CLEC4C | |
| A synthetic peptide corresponding to human CLEC4C. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only