Learn More
CPEB1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of CPEB1.
Brand: Abnova PAB6644.100ug
Description
This gene encodes a member of the cytoplasmic polyadenylation element (CPE) binding protein family. This highly conserved protein binds to a specific RNA sequence called the CPE found in the 3' UTR of some mRNAs. Similar proteins in Xenopus and mouse function to induce cytoplasmic polyadenylation of dormant mRNAs with short polyA tails, resulting in their translation. Members of this protein family regulate translation of cyclin B1 during embryonic cell divisions. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: C-PLMRNQKNRDSSSpecifications
| CPEB1 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:128000) Western Blot (0.5-2 ug/mL) The optimal working dilution should be determined by the end user. | |
| CPEB1 | |
| CEBP/CPE-BP1/CPEB/FLJ13203/MGC34136/MGC60106 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 64506 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of CPEB1. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_085097 | |
| CPEB1 | |
| A synthetic peptide corresponding to human CPEB1. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only