Learn More
CSNK1E, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of CSNK1E.
Brand: Abnova PAB7264.100ug
Description
The protein encoded by this gene is a serine/threonine protein kinase and a member of the casein kinase I protein family, whose members have been implicated in the control of cytoplasmic and nuclear processes, including DNA replication and repair. The encoded protein is found in the cytoplasm as a monomer and can phosphorylate a variety of proteins, including itself. This protein has been shown to phosphorylate period, a circadian rhythm protein. Two transcript variants encoding the same protein have been found for this gene. [provided by RefSeq
Sequence: C-PASQTSVPFDHLGKSpecifications
| CSNK1E | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:32000) Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| CSNK1E | |
| HCKIE/MGC10398 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 1454 | |
| Human | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of CSNK1E. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_001885.1;NP_689407.1 | |
| CSNK1E | |
| A synthetic peptide corresponding to human CSNK1E. | |
| 100 μg | |
| Primary | |
| Both reported variants represent identical protein (NP_001885.1 and NP_689407.1). | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only