Learn More
CYP24A1, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of CYP24A1.
Brand: Abnova PAB18975.100ug
Description
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This mitochondrial protein initiates the degradation of 1,25-dihydroxyvitamin D3, the physiologically active form of vitamin D3, by hydroxylation of the side chain. In regulating the level of vitamin D3, this enzyme plays a role in calcium homeostasis and the vitamin D endocrine system. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: C-RKYDIQATDNSpecifications
| CYP24A1 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:32000) Western Blot (0.1-0.3 ug/mL) The optimal working dilution should be determined by the end user. | |
| CYP24A1 | |
| CP24/CYP24/MGC126273/MGC126274/P450-CC24 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 1591 | |
| Human, Rat | |
| Liquid |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of CYP24A1. | |
| In 0.5mg/mL in Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_000773.2;NP_001122387.1 | |
| CYP24A1 | |
| A synthetic peptide corresponding to amino acids at internal region of human CYP24A1. | |
| 100 μg | |
| Primary | |
| This antibody is expected to recognize both reported isoforms (NP_000773.2; NP_001122387.1). | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only