missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyp2d5, Goat, Polyclonal Antibody, Abnova™
Description
Sequence: C-DAFTAQNGRSINPK
Specifications
Specifications
| Antigen | Cyp2d5 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Description | Goat polyclonal antibody raised against synthetic peptide of Cyp2d5. |
| Dilution | ELISA (1:32000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. |
| Formulation | In 0.5mg/mL Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) |
| Gene | Cyp2d5 |
| Gene Accession No. | NP_775426.1 |
| Gene Alias | Cyp2d10/MGC108540 |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?