missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DEFA1/DEFA3, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of DEFA1/DEFA3.
Brand: Abnova PAB18743.100ug
This item is not returnable.
View return policy
Description
Sequence: C-DESLAPKHPGSRKNSpecifications
| DEFA1/DEFA3 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:128000) Western Blot (1-3 ug/mL) Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (4-6 ug/mL) The optimal working dilution should be determined by the end user. | |
| NP_004075.1 (Gene ID : 1667);NP_005208.1 (Gene ID : 1668) | |
| DEFA1 | |
| A synthetic peptide corresponding to internal region of human DEFA1/DEFA3. | |
| 100 μg | |
| Primary | |
| This antibody is expected to equally recognize DEFA1 precursor and DEFA3 precursor. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of DEFA1/DEFA3. | |
| In Tris saline, pH 7.3 (0.02% sodium azide, 0.5% BSA) | |
| DEF1/DEFA2/HNP-1/HP-1/MGC138393/MRS | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 1667/1668 | |
| Human | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction