missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Donkey IgG (H+L) Secondary Antibody, Biotin, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB120-6987
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Donkey samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Specifications
| IgG (H+L) | |
| Polyclonal | |
| Western Blot 1:2000 - 1:10000, ELISA 1:20000 - 1:100000, Immunohistochemistry 1:1000 - 1:5000 | |
| Donkey IgG whole molecule | |
| 1 mg | |
| Adaptive Immunity, Immunology | |
| Prepared from monospecific antiserum by immunoaffinity chromatography using Donkey IgG coupled to agarose. Assay by immunoelectrophoresis resulted in a single precipitin arc against antibiotin, anti-Goat Serum, Donkey IgG and Donkey Serum. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Biotin | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Secondary | |
| Donkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction