missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Donkey IgG whole molecule,Polyclonal Secondary Antibody, Abnova™
Brand: Abnova PAB9194.2mL
Denna artikel kan inte returneras.
Se returpolicy
Specifikationer
| Goat anti-Donkey IgG whole molecule | |
| Polyclonal | |
| Goat anti-donkey IgG recognizes donkey IgG whole molecule. This secondary antibody was purified using delipidation and defibrination. | |
| Lyophilized from 10mM KH2PO4, 150mM NaCl, pH 7.2 (0.01% sodium azide) | |
| Donkey IgG whole molecule | |
| 2 mL | |
| Donkey | |
| Lyophilized |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| The optimal working dilution should be determined by the end user. | |
| Goat | |
| This product was prepared from monospecific antiserum by a delipidation and defibrination. Assay by immunoelectrophoresis resulted in a single precipitin arc against donkey IgG and donkey serum. | |
| Secondary | |
| Store at 4°C on dry atmosphere. After reconstitution with 2mL of deionized water store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
For Research Use Only
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering