Learn More
EBAG9, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of EBAG9.
Brand: Abnova PAB7053.100ug
Description
This gene was identified as an estrogen-responsive gene. Regulation of transcription by estrogen is mediated by estrogen receptor which binds to the estrogen-responsive element (ERE) found in the 5'-flanking region of this gene. The encoded protein is a tumor-associated antigen that is expressed at high frequency in a variety of cancers. Two transcript variants differing in the 5' UTR, but encoding the same protein, have been identified for this gene. [provided by RefSeq]
Sequence: C-KEAQRLMKKEQNSpecifications
| EBAG9 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:64000) Western Blot (1-3 ug/mL) The optimal working dilution should be determined by the end user. | |
| EBAG9 | |
| EB9/PDAF/RCAS1 | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 9166 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of EBAG9. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_004206.1 | |
| EBAG9 | |
| A synthetic peptide corresponding to human EBAG9. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only