Learn More
EIF2B4, Goat, Polyclonal Antibody, Abnova™
Goat polyclonal antibody raised against synthetic peptide of EIF2B4.
Brand: Abnova PAB19012.100ug
Description
Eukaryotic initiation factor 2B (EIF2B), which is necessary for protein synthesis, is a GTP exchange factor composed of five different subunits. The protein encoded by this gene is the fourth, or delta, subunit. Defects in this gene are a cause of leukoencephalopathy with vanishing white matter (VWM) and ovarioleukodystrophy. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: C-RKDYGSKVSLFSHSpecifications
| EIF2B4 | |
| Polyclonal | |
| Unconjugated | |
| ELISA (1:16000) Western Blot (0.3-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| EIF2B4 | |
| DKFZp586J0119/EIF-2B/EIF2B/EIF2Bdelta | |
| Goat | |
| Antigen affinity purification | |
| RUO | |
| 8890 | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| ELISA, Western Blot | |
| 0.5 mg/mL | |
| Goat polyclonal antibody raised against synthetic peptide of EIF2B4. | |
| In Tris saline, pH 7.3 (0.5% BSA, 0.02% sodium azide) | |
| NP_751945.2;NP_001029288.1;NP_056451.3 | |
| EIF2B4 | |
| A synthetic peptide corresponding to amino acids at internal region of human EIF2B4. | |
| 100 μg | |
| Primary | |
| Human | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only