missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Equine IgG Fc Secondary Antibody, PE/Cy5.5, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-72727PECY55
This item is not returnable.
View return policy
Description
IgG Fc Polyclonal antibody specifically detects IgG Fc in Equine samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| IgG Fc | |
| Polyclonal | |
| PBS | |
| Horse IgG F(c) fragment | |
| 500 μg | |
| Adaptive Immunity, Immunology | |
| Equine | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry | |
| PE-Cyanine5.5 | |
| Goat | |
| Immunogen affinity purified | |
| RUO | |
| Secondary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction